Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2X YT Agar plates with Carbenicillin, Teknova

Y4236 Y6230 Y4238 Y4235 Y4235 Y6260 Y4230 Y4391-4PK Y4265 Y4390 Y4236 Y4265 Y4238 Y4235 Y6260 Y4390 Y6230 Y4391
101320-796PK 207.12 USD InStock
101320-796 101320-840 100217-086 100219-374 100219-792 101446-776 101320-800 CA101446-776 CA101320-796 CA101320-840 CA100219-364 CA100219-792 CA100219-374 CA100219-026 100219-026 CA100217-086 CA101320-800
2X YT Agar plates with Carbenicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
These sterile pre poured agar plates are enriched with tryptone, yeast extract, sodium chloride, agar, glucose, carbenicillin

  • Shipping conditions: Ambient
  • Storage conditions: Refrigerate

They are designed for growth and propagation of recombinant E.coli and used with carbenicillin resistant strains.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty