PureClean™ self-sealing autoclave bags are made from a transparent film and 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilization applications. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.
- Clear side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
- Convenient self-seal closure eliminates the need for autoclave tape or the validation and use of a heat sealer
- Fast and consistent for equipment preparation area
- Resists tearing and punctures
- Superior microbial barrier
- Manufactured and packaged in a certified ISO 6 cleanroom
- Made in the USA
- Certificates of Conformance sent with shipment
The drapeable nature of the 1421B Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
Certifications: Certificates of conformance sent with each shipment.