Would you like to log in again?
Zymolyase Ultra, Zymo Research
Supplier: Zymo Research
E1007T E1007-10 E1007-2
77529-390EA 189.96 USD InStock
77529-390 77529-392 77529-388
Zymolyase Ultra, Zymo Research
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Zymolyase Ultra is a novel formulation of enzyme that is optimized for efficient yeast cell wall digestion.
  • Ultra efficient: 50× more efficient than lyticase
  • Ultra Low-Bioburden: Up to 70× less DNA contamination compared to other suppliers
  • Convenient to use: Dissolves in water and stable for multiple freeze-thaw cycles
Digestion of yeast and fungal cell walls is necessary for many experimental procedures. Lytic enzymes such as Zymolyase, Lyticase, Glucanase or Chitinase are routinely used for digestion of cell walls. Distinguished from conventional lytic enzymes, Zymolyase Ultra is a novel formulation of enzyme that is optimized for efficient yeast cell wall digestion. It contains enzymes that can degrade several different components of yeast cell walls. It is extensively purified using a novel DNA/RNA removal technology that results in extremely low nucleic acid contamination. It is suitable for qPCR and sequencing-based assays where low DNA/RNA contamination is critical.
Packaging: Supplied in tubes/capped bottles
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty