Would you like to log in again?
CELLCOAT® SCREENSTAR MultiCoat Test Plates
Supplier: Greiner bio-one
CELLCOAT® SCREENSTAR
655866-MCP 781866-MCP
77529-446CS 738.23 USD InStock
77529-446 77529-448
CELLCOAT® SCREENSTAR MultiCoat Test Plates
Microplates Cell Culture Plates
Test plates designed to help users find the optimal cell culture surface for assays.
  • Offers 11 different peptide and protein coatings on a single microplate to test various biological surfaces simultaneously
  • Equipped with a cycloolefin bottom that has glass-like optical properties and planarity for optimal image quality
  • Available in 96-well and 384-well formats to accommodate different assay needs and throughput requirements
  • Improves cell attachment, spreading, distribution, and growth, and can modulate differentiation and protein expression
  • Allows for quick and straightforward testing of multiple biological coatings to find the most suitable in-vivo mimicking environment for specific cells
The multiple biomolecular coatings enhance binding capacity and specificity, making multicoat plates ideal for diverse high-throughput applications. Both 96- and 384-well formats are compatible with automated systems, ensuring seamless integration into laboratory workflows.
Ordering information: Ask about black CELLCOAT® SCREENSTAR plates with cycloolefin available in 96, 384, and 1536-well formats.
Packaging: Packaged 2 pieces per bag (plate and lid).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty