Would you like to log in again?
Personna® Utility Blades
Supplier: AccuTec Blades
Personna®
61-0849-0000
77529-508BX 59.24 USD InStock
77529-508
Personna® Utility Blades
Razors and Blades
Personna Blue Armor Edge™ utility blades are made with premium steel using a proprietary grinding process for the best balance of sharpness and durability.
  • 0.025" heavy duty carbon steel
  • Multi-purpose blade for various cutting jobs
  • Fits most standard 3-notch compatible utility knives
Each carton includes 100 blades packaged in tubes of 10 blades each.
Packaging: 10 blades per BladeGard dispenser.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty