Would you like to log in again?
AccuTec Pro Single Edge Blades, Individually Wrapped
Supplier: AccuTec Blades
AccuTec Pro
APBL-7054-0000 APBL-7065-DISP
77529-512BX 11.61 USD InStock
77529-512 77529-514
AccuTec Pro Single Edge Blades, Individually Wrapped
Razors and Blades
AccuForge single edge blades feature two-facet edges for durability. Blades are wrapped in protective paper.
  • Two-facet edge for durability
  • Available in 0.009" regular duty or 0.012" heavy duty blades
  • Re-closable clamshell for safe and convenient blade storage
Each clamshell contains 100 blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty