Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cole-Parmer® RV-200 Rotary Evaporators

Supplier: Cole Parmer
Cole-Parmer®
52411-94 52411-92
77529-542EA 2236.85 USD InStock
77529-542 77529-544
Cole-Parmer® RV-200 Rotary Evaporators
Evaporators Rotary Evaporators
Simple one-handed lift operation for ease of use.

  • Set and view rotation speed, temperature, and timer on two large LCDs
  • PTFE sealing system offers excellent thermostability
  • PID temperature controller provides stable temperature control
  • Small footprint takes up minimal laboratory space

Easily set and view operating parameters on the two large digital displays; one for rotation speed and timer and another for bath temperature and timer. The PTFE sealing system provides great thermostability and excellent chemical resistance.

A stainless steel bath with PTFE coating withstands high temperatures and is easy to clean. Safety features include dry-run protection, overtemperature shutoff, circuit breaker and power interruption alarm.

Speed resolution: 1 rpm
Glass type: Uncoated
Temperature resolution: 0.1 °C
Vacuum seal: PTFE
Vacuum connection: Angled 10 mm GL 14 barb
Temperature safety limiter: Yes

Delivery information: Supplied with evaporator control stand, glassware set* (Includes a 1 L evaporation flask but accommodates either a 1 or 2 L flask), water bath, clip, vacuum gauge, and power cords with NEMA 5 plug for rotator and bath.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty