Would you like to log in again?
VersaHOCl™ Sterile Disinfectant Cleaner
Supplier: Berkshire
VersaHOCl™
VHOCL6QST
77530-434CS 192 USD InStock
77530-434
VersaHOCl™ Sterile Disinfectant Cleaner
Cleaning Agents Disinfectants
VersaHOCl™ Sterile is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, Berkshire’s VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.
  • Kills common microbiological contaminations on surfaces
  • Can be used in a variety of applications
  • Sterile filled and provided double bagged for easy transfer into critical environments
  • Product certificate provided with each case
  • Ideal for large surface cleaning
  • No strong odor provides good user acceptability
  • No special disposal is required
  • Facilitates transfer disinfection into cleanroom
  • No mixing required
Non-toxic with low odor and low residue, VersaHOCl™ establishes the safest standards in disinfectant technology. With an active ingredient of 0.065% hypochlorous acid, it’s non-corrosive and effective on hard, non-porous surfaces. This versatile disinfectant is ideal for use in controlled environments such as labs, cleanrooms, compounding pharmacies, and manufacturing facilities. It can also be used in areas such as hospitals, clinics, healthcare facilities, schools, and government and office buildings.
Certifications: EPA registration number: 95747192998. CA Dept. of Pesticides Regulation Number: 95737-1-AA-92998.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty