Would you like to log in again?
Micro-Touch® DermaClean® Examination Gloves, Latex
Supplier: Ansell
Micro-Touch® DermaClean®
107427 107428 107425 107426 107429
77531-238BX 10.8 USD InStock
77531-238 77531-234 77531-236 77531-230 77531-232
Micro-Touch® DermaClean® Examination Gloves, Latex
Gloves
These ambidextrous gloves are fully textured and therefore provide an excellent grip in wet and dry conditions. They are ideal for instrumentation, handling test tubes or infusion equipment in labs, pharmaceutical environments and general wards.
  • Latex, powder-free medical
  • For an excellent grip in wet and dry conditions
  • For easy donning
  • Provides great comfort and flexibility
  • Secure grip
  • Comfortable fit and easy donning
  • Freedom from holes: 1,5 AQL
Recommended for brief, low-risk examination procedures.
Certifications: Certified of EN ISO 21420:2020, ISO 10993-1, ISO 11193-1:2008, ISO 10993-5 and ISO 10993-10:2002. ISO 9001, ISO 13485 and ISO 10993-10.
Caution: This product contains natural rubber latex, which may cause allergic reactions.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty