Would you like to log in again?
ValuSeal-HA® Sealed-Edge Cleanroom Wipers, Berkshire
Supplier: Berkshire
ValuSeal-HA®
VSHA.0909B.8 VSHA1212B14 VSHA.0909.8 VSHA.1212.14
80094-518CS 945.14 USD InStock
80094-518 77531-240 80094-522 80094-520
ValuSeal-HA® Sealed-Edge Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Highly absorbent, 100% knitted polyester wipers have ultrasonic heat-sealed edges.
  • Lightweight
  • Low particles, fibers, ions, and extractable
  • High absorbency material reduces actual cost in use compared to heavier weight products
Wipers work well with IPA, acetone, and other solvents. Steam autoclavable for aseptic environments.
Certifications: Recommended for ISO Class 3 (FED-STD-209E Class 1/M1.5) cleanroom environments, including semiconductor and biomedical cleanroom applications.
Ordering information: Wipers are laundered and packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty