Would you like to log in again?
Drop In Sink, Self Rim Design with No Deck, One Bowl
Supplier: Eagle MHC
SR16-19-13.5-1-ND SR21-15.75-10-1-ND SR16-13.5-10-1-ND SR22-22-13.5-1-ND SR28-16-11.5-1-ND SR10-14-9.5-1-ND SR14-16-9.5-1-ND SR24-24-13.5-1-ND SR10-14-5-1-ND SR16-19-8-1-ND SR18-24-13.5-1-ND
77531-248EA 2498.14 USD InStock
77531-248 77531-256 77531-244 77531-258 77531-246 77531-252 77531-262 77531-254 77531-242 77531-250 77531-260
Drop In Sink, Self Rim Design with No Deck, One Bowl
Furniture Casework
Sinks can be mounted onto top or bottom of countertop. Heavy gauge type 300 series stain less steel coved bowls with large radius.
  • All sinks feature 35 mm (¹³/₈") trim on all four sides of sink
  • 121 mm (⁴³/₄") sump in sink bowl
  • 89 mm (³¹/₂") basket drain with 38 mm (¹¹/₂") outlet
  • Self rimming
16, 18 or 20 gauge industrial grade construction and quality. 16 gauge bowls are fabricated, 18 and 20 gauge bowls are deep-drawn.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty