Would you like to log in again?
Peroxigard™ Concentrate
Supplier: Lighthouse
Peroxigard™
PCON242311 PCON242305 PCON242309
77532-202EA 229.71 USD InStock
77532-202 77532-206 77532-204
Peroxigard™ Concentrate
Cleaning Agents Detergents and Surface Cleansers
One-step disinfectant cleaner and deodorizer.
  • Cleans and achieves disinfection contact time compliance against pathogens of concern in one step
  • Two times faster than leading concentrate disinfectant products
  • Designed with animal safety in mind
  • Fragrance free, non-irritating, non-corrosive, and superior metal compatibility
  • Eliminates multi-product programs and activation time, improving productivity and compliance
Peroxigard™ concentrate utilizes accelerated hydrogen Peroxide®, a patented and synergistic blend of commonly used ingredients, that when combined with low levels of hydrogen peroxide dramatically increase its efficacy and cleaning performance. Peroxigard™ concentrate is a simple and effective way to clean and disinfect floors, ceilings, and walls within the life science research setting. This revolutionary technology streamlines processes with no multi-part mixing and no activation. This concentrate has a long shelf life once diluted, to aid in reducing waste.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty