Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® STAT 4 Clinical Centrifuge

Supplier: Avantor
VWR®
00-053-037-000
77553-262EA 3666 USD InStock
77553-262
VWR® STAT 4 Clinical Centrifuge
Centrifuges Benchtop Centrifuges
VWR STAT clinical benchtop centrifuges spin samples in 5 minutes or less for the most urgent chemistry and haematology testing.

  • 3 convenient presets for common STAT cycles, select using simple two-button interface
  • LED lid lighting system indicates cycle status: Off when ready, on when running, flashing when done
  • Max speed 8500 min⁻¹ (rpm)/4400×g
  • 4 tube capacity (75 mm/6 ml)
  • Compact centrifuge (7×7×7") to fit with minimal bench space
  • Fixed angle rotor design for optimal processing of coag and platelet poor plasma
  • 2 year warranty

The VWR STAT 4 is a high-speed benchtop centrifuge designed to rapidly separate fluids in a syringe-type collection tube. Its compact size saves on bench space and allows samples to be moved easily around the laboratory. The simple two-button interface makes it easy to select a preset or program custom STAT cycles based on your IFUs and unique validation needs. Use all three presets, or set and lock to a single cycle for maximum repeatability. Enjoy years of worry-free operation with the VWR STAT 4’s brushless motor, reinforced carbon fibre components, and a two-year calibration guarantee. Enhance lab safety with imbalance protection and a shatterproof lid, which locks until the centrifuge has stopped completely.

Certifications: MET, UKCA.

Delivery information: Each centrifuge is supplied ready to run with rotor, tube holders, QR code manual and quick start guide.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty