Would you like to log in again?
VWR® STAT Coag Clinical Centrifuge
Supplier: Avantor
VWR®
00-676-037-001
77553-266EA 4009.82 USD InStock
77553-266
VWR® STAT Coag Clinical Centrifuge
Centrifuges Benchtop Centrifuges
VWR STAT clinical benchtop centrifuges spin samples in 5 minutes or less for the most urgent chemistry and haematology testing.
  • 3 convenient presets for common STAT cycles, select using simple two-button interface
  • LED lid lighting system indicates cycle status: Off when ready, on when running, flashing when done
  • Max speed 6600 min⁻¹ (rpm)/4400×g
  • 12 tube capacity (100 mm /10 ml)
  • Compact size to be used in an array in accessioning, further driving down TAT
  • Fixed angle rotor design for optimal sample processing
  • 2 year warranty
Optimise for PPP with the VWR® STAT Coag Clinical Centrifuge. Designed with platelet poor plasma spins in mind, the VWR® STAT Coag offers three convenient presets for common laboratory needs. Its fixed angle rotor spins up to 12×100 mm/10 ml tubes at a time. With a compact footprint and simple two-button controls, the VWR® STAT Coag is right at home in accessioning, or for tube processing in the emergency department. Max speed 4400×g/6600 min⁻¹ (rpm).
Certifications: CE, MET, UKCA
Delivery information: Each centrifuge is supplied ready to run with rotor, tube holders, QR code manual and quick start guide. Microtainer tube adapters must be ordered separately.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty