Would you like to log in again?
PAXgene™ Blood RNA Tube, BD Biosciences
Supplier: BD Biosciences
PAXgene™
762165
77776-026CS 2239.05 USD InStock
77776-026
PAXgene™ Blood RNA Tube, BD Biosciences
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The PAXgene™ Blood RNA tube is part of a system that standardizes and integrates the key steps of whole blood collection, nucleic acid stabilization, and RNA purification
By minimizing the unpredictability of RNA processing, the system provides increased accuracy of intracellular RNA analysis. Tubes contain a proprietary reagent that immediately stabilizes intracellular RNA for three days at 18 to 25°C (64 to 77°F) and five days at 2 to 8°C (35 to 46°F). The ability to standardize the preanalytical process greatly enhances test accuracy and lab efficiency. Tube size is 16x100mm, with a capacity of 2.5 mL. Tubes feature a clear BD Hemogard™ closure and paper label for marking.
Certifications: CE marked.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty