Would you like to log in again?
Adapters, Pressure Vessel, Chemglass
Supplier: Chemglass
CG-1881-01 CG-1881-02
77776-448EA 119.93 USD InStock
77776-448 77776-446
Adapters, Pressure Vessel, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapter permits rotary evaporation of the contents of any pressure vessels after the reaction is complete.
  • Eliminates the need to transfer samples into a conventional round bottom flask
  • Joint sizes other than 24/40 are available
  • Bushing and o-ring included
Adapter can also be connected to any vacuum line having a 24/40 port or used in conjunction with a adapter to connect to hoses.
Delivery information: Supplied complete with one open top bushing and Viton o-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty