Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PYREX® Petri Dishes, Corning
Supplier: Corning
PYREX®
3160-150 3160-60 3160-150BO 3160-152BO 3160-101CO 3160-102CO 3160-152CO 3160-60CO 3160-102BO 3160-101BO 3160-100BO 3160-100CO 3160-101 3160-150CO 3160-100 3160-60BO 3160-152 3160-102
77777-028CS 89.87 USD InStock
77777-028 25354-105 25354-127 25354-025 25354-069 25354-047 25356-042 77777-030 25354-080 77777-032 33500-292 77777-034 77777-022 77777-036 33500-296 77777-024 77777-026 33500-294
PYREX® Petri Dishes, Corning
Culture Dishes Petri Dishes
These dishes withstand repeated wet and dry sterilization.
  • Beaded Edges for Greater Mechanical Strength
  • Color-Coded Tops and Bottoms
  • Triangular Enamel Reference Points
  • Dishes Withstand Wet and Dry Sterilization
Edges are beaded to provide greater mechanical strength, to equally space bottom and side walls, and to reduce capillary action of condensed moisture on the sides.
For easy sorting, tops are blue, and bottoms are white. Bottoms feature a triangular enamel reference point for serial dilutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty