Would you like to log in again?
BCR® Cleanroom Bond Paper, Berkshire
Supplier: Berkshire
BCR®
BB104.0811.10W BB85081110B BB85081110W BB85081110G BB6808118B BB85081110P BB1041117.5W BB1041117.5B BB85081110C BB104081110W BB104081110B BB6808118W
76238-470CS 444.81 USD InStock
76238-470 77788-006 77788-018 77788-008 77788-002 77788-024 77788-014 77788-036 77788-016 77788-038 77788-010 77788-012
BCR® Cleanroom Bond Paper, Berkshire
Paper
Designed to minimize contamination and run through copiers, laser printers, and impact printers.
  • Purest cellulose to minimize contamination
  • Specialized coating to reduce particles, fibers, and micro-organisms
  • Retains toner from copiers and laser printers to reduce smudging
  • Autoclavable
  • Latex-free
  • ISO Class 3–8
  • Cleanroom Class 1–100,000
Optimal strength and durability ensures maximum consistency in printing and photocopying. A cellulose base sheet with a synthetic latex binder is added during manufacturing to reduce the potential of particle or fiber contamination. Paper features a fiber coating that enhances strength and durability without limiting the ability to absorb inks or bind toner. Coating is resistant to the heat and static normally generated in photocopying and laser printing. Consult printer manual before using any kind of paper.
Can also be used as pages in cleanroom manuals and as lot tracer sheets or batch records in pharmaceutical manufacturing areas. Cleanroom compatible labels on the end of packs for easy identification on shelves.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty