Would you like to log in again?
Rotors for Centrifuges 5804(R) and 5810®
Supplier: Eppendorf
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5820772000 022638505 5820765003 5804746007 022637461 5820760001 022637100 022637207 022638581 022638564 5820771003 5804746007 022637401 5820715006 022638581 5820771003 5820765003 5820760001 5820772000 022637002
53574-220EA 4086.11 USD InStock
CA11017-462 CA11017-452 CA11017-454 53574-220 47743-188 77888-214 77888-208 CA47745-380 53513-820 53513-840 89305-270 89305-274 53513-818 89305-240 CA11017-458 89305-264 53513-816 89305-266 CA11017-428 97058-952
Rotors for Centrifuges 5804(R) and 5810®
Centrifuges Rotors
Eppendorf rotors for centrifuges 5804(R) and 5810(R) precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorise with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight options available
Experience precision in high-performance centrifugation with Eppendorf Rotors designed for centrifuge 5804/5804 R. These rotors offer diverse capacities and speeds, providing flexibility for various sample processing needs. Accessorise with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-performance applications, the centrifuge 5804 series is further optimised with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your centrifugation experience.
Ordering information: Rotors, adapters, and accessories are only for use with centrifuge models specified. Unless otherwise specified, rotors may be used with all centrifuge models.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty