Would you like to log in again?
FILTER RIGID MERV15 20X24X12
Supplier: Total Filtration Services
Filter Rigid Merv15 20X20X12^ Lg-913 90[perCent]-95[perCent], unique channel flow molded media, merv 11 and 15, very low resistance, high dust holding capacity, all plastic construction, sustainable component for leed/green building initiative, exceeds leed/green merv 13 requirement, lower energy cost.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER RIGID MERV15 20X24X12 78003-288 LG915 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty