Would you like to log in again?
Protease Inhibitor Cocktail Set I, Animal-Free, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
Calbiochem®
535142-1SET 535142-1ML 535142-1SET 535142-1ML
80017-340EA 71.91 USD InStock
CA80017-342 CA80017-340 80017-340 80017-342
Protease Inhibitor Cocktail Set I, Animal-Free, Calbiochem®, MilliporeSigma
Cell Culture Supplements and Additives
Cocktail of five protease inhibitors for the inhibition of a broad range of proteases and esterases
Lyophilized. Reconstitute each vial with 1 mL dH2O to yield a 100X stock solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty