Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HMS174 Competent Cells, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
69452-3 69454-4 69453-4 70234-3 69453-3 69454-3 69453-3 69454-4 69453-4 69452-4 69452-3 69452-4
80030-334EA 245.11 USD InStock
80030-334 CA80030-332 80030-336 80030-832 CA80030-334 CA80030-340 80030-330 80030-332 CA80030-330 80030-340 80030-338 CA80030-336
HMS174 Competent Cells, Expression Strains, MilliporeSigma
Competent Cells
A common method for transformation of plasmids into E.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lac UV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
The standard volume per tube is 0.2 ml, which provides enough cells for 10 transformations. HMS174 Competent Cell kits are supplied with 2×0.2 ml, SOC medium and test plasmid. Several strains are also available as Singles™ Competent Cells, which are provided in single-use, 50 μl volumes for extra convenience and efficiency.
Coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen® prepared competent cells. Every Novagen® competent cell strain is verified for phenotype and purity and is guaranteed for transformation efficiency.
HMS174 strains provide the recA mutation in a K-12 background. Like BLR, these strains may stabilize certain target genes whose products may cause the loss of the DE3 prophage.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty