Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BLR Competent Cells, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
Novagen®
69053-3 69956-4 69956-3 69053-4 69053-3 69956-4 69053-4 69956-3
80030-584EA 289.54 USD InStock
CA80030-582 80030-584 CA80030-584 CA80030-082 80030-582 80030-080 80030-082 CA80030-080
BLR Competent Cells, Expression Strains, MilliporeSigma
Competent Cells
A common method for transformation of plasmids into E. coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen® prepared competent cells. Every Novagen® competent cell strain is verified for phenotype and purity, and is guaranteed for transformation efficiency.
  • Guaranteed transformation efficiencies of >2×10⁶ cfu/µg
  • Deficient in Lon and OmpT proteases
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. BLR is a recA- derivative of BL21 that may help stabilise target plasmids containing repetitive sequences or whose products may cause the loss of the DE3 prophage (Roca, Studier 1991).
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilising pET recombinants encoding target proteins that affect cell growth and viability.
The standard volume per tube is 0.2 ml, which provides enough cells for 10 transformations. All kits are supplied with SOC medium and test plasmid. Several strains are also available as Singles™ Competent Cells, which are provided in single-use, 50 μl volumes for extra convenience and efficiency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty