Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Singles™ Competent Cells, Novagen®
Supplier: Novagene
Singles™
70181-4 70236-4 70236-3 70236-3 70181-3 70235-3 70236-4 70181-4 70235-4 70181-3 70235-4
80030-840EA 584.02 USD InStock
80030-840 80030-752 80030-754 CA80030-752 CA80030-840 CA80030-836 80030-838 CA80030-838 CA80030-754 80030-834 80030-836
Singles™ Competent Cells, Novagen®
Competent Cells
Designed for convenience and reliability for high efficiency plasmid transformation
NovaBlue—Highly recommended for general cloning purposes. Features extremely high transformation efficiency, blue/white screening of recombinants by lacZ [alpha]-complementation, endA and recA mutations for high plasmid quality and yield, and F factor to support M13 growth and single stranded plasmid rescue of vectors carrying the f1 origin of replication.
BL21(DE3) and BL21(DE3)pLysS—Strains are widely used hosts for protein expression from pET recombinants. Suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. BL21 strains are deficient in both lon and ompT proteases. Designation of DE3 indicates that the host is a lysogen of [lambda]DE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Strains carrying pLysS express T7 lysozyme. This lysozome further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
For carbenicillin, an antibiotic less sensitive to low pH than ampicillin, see 80030-956.
Cells are provided in 50µL single-use aliquots that eliminate the need to sub-aliquot, freeze or thaw, or waste partially used vials. Transformation can be done in the tube containing the cells. Two types of host strains are available: NovaBlue and BL21(DE3)/BL21(DE3)pLysS.
Ordering information: Cells are provided as frozen, single-use aliquots in 11-reaction or 22-reaction kits that include Test Plasmid and SOC Medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty