Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Perfect DNA™ Markers, MilliporeSigma
Supplier: MilliporeSigma
70087-3 70087-3 69002-3 69002-3 70540-3 70540-3
80030-890EA 225.29 USD InStock
CA80030-890 80030-890 CA80031-040 80030-932 80031-040 CA80030-932
Perfect DNA™ Markers, MilliporeSigma
Electrophoresis Reagents Molecular Weight Markers
Easy-to-remember sizes for routine use in plasmid cloning.
  • Easy-to-remember sizes
  • Uniform band intensities
  • Instant band identification
The Perfect DNA™ Markers contain sets of DNA species having convenient, easy-to-remember sizes for agarose gel analysis. The markers have uniform band intensities except for the easily identifiable reference bands, which are useful for instant band identification.
Delivery information: An extra vial of 6X DNA Gel Loading Buffer is included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty