Would you like to log in again?
Caspase-6 Substrate III, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
Calbiochem®
218763-5MG 218763-5MG
80050-850EA 316.29 USD InStock
CA80050-850 80050-850
Caspase-6 Substrate III, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents
A fluorogenic substrate for caspase-6 (Mch2)
Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Excitation maximum: ~400nm; emission maximum: ~505nm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty