Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Caspase-3 Substrate IV, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
Calbiochem®
264150-1MG 264150-1MG 264150-5MG 264150-5MG
80051-370EA 409.56 USD InStock
CA80051-370 80051-370 80051-368 CA80051-368
Caspase-3 Substrate IV, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents
Fluorogenic caspase-3 substrate
Also acts as a substrate for granzyme B-processed ICE-LAP3. Reaction can be monitored by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Also a substrate for caspase-6, caspase-7, caspase-8, and caspase-10.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty