Would you like to log in again?
beta-Glucosidase
Supplier: EMD Millipore
Calbiochem®
345788-5MG
80051-862EA 354.69 USD InStock
80051-862
beta-Glucosidase
Enzymes
β-Galactosidase, E. coli an inducible enzyme not found in eukaryotic cells that is used as a reporter gene.
  • Unit of definition: One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol of p-nitrophenyl-β-galactoside per minute at 37 °C
  • Specific activity: ≥600 units/mg protein
Used to assess efficiency of transfection. Also used in ELISA and immunocytochemical techniques. Histochemical substrates include X-Gal.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty