Would you like to log in again?
Human Myeloperoxidase (from Polymorphonuclear Leukocytes)
Supplier: MilliporeSigma
475911-100UG 475911-100UG
80053-242EA 237.8 USD InStock
80053-242 CA80053-242
Human Myeloperoxidase (from Polymorphonuclear Leukocytes)
Enzymes
100mM NaCl, 50mM sodium acetate, pH 6.0
A lysosomal heme protein that catalyzes oxidations by hydrogen peroxide, including MPO-chloride-mediated killing of microbes and tumor cells, inactivation of chemotactic factors and cross-linking of proteins. Specific activity: 150–200units/mg protein. Prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty