Would you like to log in again?
Phospholipase D, Streptomyces chromofuscus, Enzyme Assay, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
525200-250U
80053-704EA 319.28 USD InStock
80053-704
Phospholipase D, Streptomyces chromofuscus, Enzyme Assay, MilliporeSigma
Assays Enzyme Assays
Native phospholipase D from Streptomyces chromofuscus
Catalyzes the hydrolysis of lecithin to phosphatidic acid and choline. Coupled with choline oxidase, it can also be used for the determination of phospholipids in serum.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty