Would you like to log in again?
Rabbit 20S Proteasome
Supplier: EMD Millipore
539154-10UG
80053-870EA 461.74 USD InStock
80053-870
Rabbit 20S Proteasome
Enzymes
Catalytic core enzyme of the 26S proteasome that degrades ubiquitinated proteins
Biological activity: fully functional as measured using Z-LLVY-AMC as the substrate following SDS activation.
Hydrolyzes various peptide substrates and proteins with broad specificity in a non-ATP-dependent process. Can be activated by PA28 (80053-556) or SDS.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty