Would you like to log in again?
FeTPPS
Supplier: EMD Millipore
341492-25MG 341492-25MG
80056-448EA 110.88 USD InStock
CA80056-448 80056-448
FeTPPS
Enzymes
A ferric porphyrin complex that catalytically isomerizes peroxynitrite to nitrate both in vivo and in vitro.
Provides cytoprotection against peroxynitrite (EC50=5 µM). Does not complex with nitric oxide and exhibits minimal SOD mimetic activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty