Would you like to log in again?
MCl-186, Calbiochem
Supplier: Calbiochem
443300-500MG
80056-994EA 96.66 USD InStock
CA80056-994 80056-994
MCl-186, Calbiochem
Cell Culture Supplements and Additives
A free radical scavenger and antioxidant that reduces post-ischemic brain injury
Inhibits iron-dependent peroxidation in rat brain homogenates (IC50=15µM) and mitochondrial homogenates (IC50=2.3µM).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty