Would you like to log in again?
Melatonin, Calbiochem
Supplier: Calbiochem
444300-1GM 444300-1GM
80057-004EA 101.27 USD InStock
80057-004 CA80057-004
Melatonin, Calbiochem
Cell Culture Supplements and Additives
A hormone that has been postulated as the mediator of photic-induced anti-gonadotropic activity in photoperiodic mammals. Counteracts the apoptotic effects of etoposide in bone marrow cells. Melatonin receptors are coupled to a G-protein system. Inhibits rat cerebellar nitric oxide synthase (NOS). Also acts as a peroxynitrite scavenger.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty