Would you like to log in again?
Xanthine Oxidase (from Buttermilk)
Supplier: EMD Millipore
Calbiochem®
682151-10U 682151-50U
80057-716EA 312.86 USD InStock
80057-716 80057-714
Xanthine Oxidase (from Buttermilk)
Enzymes
Native xanthine oxidase from buttermilk
Complex metalloprotein that catalyzes oxidative hydroxylation of a variety of aromatic heterocycles and simple aldehydes. Catalyzes the oxidation of xanthine to uric acid.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty