Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HEPES, Free Acid Solutions, ULTROL® Grade, Calbiochem®, MilliporeSigma

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

375368-500ML 375368-500ML 375368-100ML 375368-100ML
80058-194EA 491.49 USD InStock
CA80058-194 80058-194 CA80058-192 80058-192
HEPES, Free Acid Solutions, ULTROL® Grade, Calbiochem®, MilliporeSigma
Buffers Cell Culture Buffers
A zwitterionic N-substituted amino sulfonic acid buffers active in the pH range of 6.0 to 8.5.

  • Absorbance: <0.05 at 260 nm, 1 M, H₂O
  • Sterile filtered
  • pKa: 7.55 at 20 °C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty