Would you like to log in again?
HEPES, Free Acid Solutions, ULTROL® Grade, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
375368-500ML 375368-500ML 375368-100ML 375368-100ML
80058-194EA 491.49 USD InStock
CA80058-194 80058-194 CA80058-192 80058-192
HEPES, Free Acid Solutions, ULTROL® Grade, Calbiochem®, MilliporeSigma
Buffers Cell Culture Buffers
A zwitterionic N-substituted amino sulfonic acid buffers active in the pH range of 6.0 to 8.5.
  • Absorbance: <0.05 at 260 nm, 1 M, H₂O
  • Sterile filtered
  • pKa: 7.55 at 20 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty