Would you like to log in again?
ULTROL® Grade, Tris Base Buffer, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
Calbiochem®
648311-5KG 648311-5KG 648311-1KG
80058-768EA 216.46 USD InStock
80058-768 CA80058-772 80058-772
ULTROL® Grade, Tris Base Buffer, Calbiochem®, MilliporeSigma
Buffers Biological Buffers
Tris Base buffer is useful in the pH range of 10.3 to 11.3 and has a pKa of 8.1 at 25 °C.
Has an absorbance <0.05 (1.0 M, H₂O, 260 nm).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty