Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ZWITTERGENT® 3-12 Detergent, Calbiochem®, MilliporeSigma

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Calbiochem®
693015-5GM 693015-5GM 693015-25GM 693015-25GM
80058-840EA 213.92 USD InStock
CA80058-840 80058-840 CA80058-838 80058-838
ZWITTERGENT® 3-12 Detergent, Calbiochem®, MilliporeSigma
Cleaning Agents Detergents and Surface Cleansers
This synthetic zwitterionic detergent retains its zwitterionic character over a broad pH range.

  • Synonyms: n-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
  • Form: White solid
  • Storage: +15°C to +30°C
  • Toxicity: Corrosive
  • CMC: 2 - 4 mM
  • Aggregation Number: 55

The zwitterionic character is attributed to the presence of both a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding to either anionic or cationic compounds.

Caution: Following reconstitution, refrigerate (4 °C). Stock solutions are stable for up to 1 month at 4 °C.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty