Would you like to log in again?
Cajon® Ultra-Torr Adapters, Chemglass
Supplier: Chemglass
AF-0066-14 AF-0066-10 AF-0066-17 AF-0066-13 AF-0066-16 AF-0066-11 AF-0066-09 AF-0066-12
80060-006EA 500.64 USD InStock
80060-006 80060-004 80060-014 80060-008 80060-002 80060-012 80060-000 80060-010
Cajon® Ultra-Torr Adapters, Chemglass
Tubing Transfer Tubing
Cajon® adapters provide the flexibility of rubber tubing.
  • Connections compensate for misalignment, expansion and contraction
  • Compressible by at least 20%
  • Extendable by 50% of its nominal listed length
Adapters are the ideal replacement for rubber, plastic, or glass tubing in critical vacuum applications.
Ordering information: Adapters are supplied complete with two AF-0067 unions, two AF-0068 cuff adapters and listed length of flexible tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty