Would you like to log in again?
Cajon Unions, AIRFREE®, Schlenk, Chemglass Inc
Supplier: Chemglass
AIRFREE®
AF-0067-22 AF-0067-06 AF-0067-21 AF-0067-20 AF-0067-05 AF-0067-04
80060-016EA 105.86 USD InStock
80060-016 80060-026 80060-018 80060-020 80060-024 80060-022
Cajon Unions, AIRFREE®, Schlenk, Chemglass Inc
Tubing Transfer Tubing
Unions are made from 316 stainless steel.
It provide seals directly from the listed sizes on a glass to glass basis, or it can be used with the cuffs and flexible tubing. They can be used with glass tubing as much as 0.0312" undersize.
Ordering information: Replacement O-Rings made of 70 durometer viton is available and must be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty