Would you like to log in again?
Airfree® Schlenk Cajon® Cuffs, Chemglass
Supplier: Chemglass
Airfree®
AF-0068-03 AF-0068-02 AF-0068-01
80060-028EA 15.46 USD InStock
80060-028 80060-030 80060-032
Airfree® Schlenk Cajon® Cuffs, Chemglass
Tubing Transfer Tubing
Cuff adapters that do not require welding or brazing to tubing to insure a leak-tight seal.
  • Made of #304 stainless steel
  • MUST be used between unions and flexible tubing
  • Leak-tight seal
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty