Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Vacuum Manifold, Chemglass
Supplier: Chemglass
AF-0451-01
80060-122EA 7228.9 USD InStock
80060-122
Vacuum Manifold, Chemglass
Manifolds
Vaccum lines constructed using 0.4 mm Chem-Vac™ Chem-Cap® valves, has 4 ports with greased glass stopcocks.
The end cap on the vacuum line is a o-ring joint No.30 while the inert gas line has a o-ring joint No.9 instead. All gauging ports are constructed using o-ring joints No.9 and are supplied complete with adapters from the O-ring joint to 9 mm OD tubulation for use with 3/8" Cajon® fittings.
Delivery information: Complete system consists of one 4-port vacuum/inert manifold, one pre-trap, one main trap assembly, one main trap body, two No.28 S.S. pinch clamps, one No.50 S.S. pinch clamp, one No.75 S.S. pinch clamp, four No.18-L S.S. pinch clamps, one No.9 o-ring joint cap stopper, three No.9 o-ring joints and four No.112 Viton o-rings.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty