Would you like to log in again?
Airfree® Schlenk Solvent Distillation Apparatus, Chemglass
Supplier: Chemglass
Airfree®
AF-0720-02 AF-0720-04
80060-168EA 2268.61 USD InStock
80060-168 80060-172
Airfree® Schlenk Solvent Distillation Apparatus, Chemglass
Distillation Components Distillation Sets
Apparatus designed for use in distillation of material such as THF or ether.
  • Storage reservoir has a capacity of 125 ml or 500 ml
  • 0 to 4 mm Chem-Vac™ Chem-Cap® valves ensure grease-free, leakproof seals
  • Top outer joint is a 24/40 while the uptake tube is a 24/40 inner joint
Extremely efficient cold finger coil condenser has a section that is immersed into the distillate. This assures that the distillate is at room temperature instead of the distillation temperature.
Side vapor tube (A) is fitted with a 14/20 standard taper outer joint, to be used with a CG-1042-14 thermometer adapter which fits a plain stem thermometer. A short (50 mm) vigreux section is provided to enhance separation. Syringe access port (B) incorporates a 4 mm PTFE stopcock with a 14/20 outer joint for use with CG-3022 septum stopper, permitting repeated syringe punctures or changes of the septum without breaching the integrity of the solvent.
Lower 3-way valve (C) allows for solvent return to flask, take-off or storage in graduated reservoir. Delivery adapter is fitted with a 12/5 spherical joint for flexibility and a 0 to 4 mm Chem-Vac™ Chem-Cap® valve (D) for purging of the receiving vessel.
Delivery information: Delivery adapter has a 14/20 inner joint and is supplied with the complete apparatus along with a stainless steel pinch clamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty