Would you like to log in again?
Glas-Col® Series O Heating Mantles, for Cylindrical Vessels, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Glas-Col®
CG-10007-16 CG-10007-10 CG-10007-18 CG-10007-14 CG-10007-12
80060-226EA 1381.59 USD InStock
80060-226 80060-224 80060-228 80060-222 80060-230
Glas-Col® Series O Heating Mantles, for Cylindrical Vessels, Chemglass
Heaters Heating Mantles
These Glas-Col® Series O mantles are used with a round bottom cylindrical reaction vessel, featuring a Duran® flange.
  • Mantle is glass fabric
  • 115 V supply
The mantle covers the bottom and sides of the vessel, leaving enough room under the flange to safely remove the reaction vessel clamp while the mantle is in place.
Delivery information: Supplied with a separable 4 foot, 2 wire extension cord with locking connector.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty