Would you like to log in again?
Universal Inlet Adapters, PTFE, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1047-07 CG-1047-14 CG-1047-11 CG-1047-13 CG-1047-03 CG-1047-10 CG-1047-01 CG-1047-20 CG-1047-05 CG-1047-12
80060-302EA 10.82 USD InStock
80060-302 80060-300 80060-306 80060-290 80060-304 80060-294 80060-282 80060-298 80060-286 80060-296
Universal Inlet Adapters, PTFE, Chemglass
Adapters Adapters, Conical Ground Joints
Unique adapter is made entirely of PTFE and is supplied complete with a series of five PTFE ferrule-style cones.
  • Inside diameters run from 3.5-6.5 mm
  • Color-coded cones for easy identification
  • Replacement PTFE cones are available in sets of 5 or individually
These cones have inside diameters that run from 3.5 to 6.5 mm and seal against the inserted tube by means of a threaded compression screw. Cones are color-coded for easy identification.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty