Would you like to log in again?
Oil Bath Combination Packages, Chemglass
Supplier: Chemglass
CG-1100-400 CG-1100-300 CG-1100-200 CG-1100-100
80060-336EA 3323.95 USD InStock
80060-336 80060-326 80060-332 80060-340
Oil Bath Combination Packages, Chemglass
Funnels Dropping Funnels
Combined oil baths with a controller and bath oil to allow users to order all of the necessary items in one package.
Package #1 consists of one each of the following items: 100×50mm Oil Bath, Digital Temperature Controller with Thermocouple, Thermocouple Clip, and 1 Quart Bath Oil .
Package #2 consists of one each of the following items: 150×75 mm Oil Bath, Digital Temperature Controller with Thermocouple, Thermocouple Clip, and 1/2 Gallon Bath Oil .
Package #3 consists of one each of the following items: 200×100 mm Oil Bath, Digital Temperature Controller with Thermocouple, Thermocouple Clip, and 1/2 Gallon Bath Oil .
Package #4 consists of one each of the following items: 100×50 mm Oil Bath, 150×75 mm Oil Bath, 200×100 mm Oil Bath, Digital Temperature Controller with Thermocouple, Thermocouple Clip, and 1/2 Gallon Bath Oil .
Ordering information: Hot plate must be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty