Would you like to log in again?
Graduated Cylinder, Chemglass
Supplier: Chemglass
CG-1224-20 CG-1224-21
80060-652EA 100.13 USD InStock
80060-652 80060-650
Graduated Cylinder, Chemglass
Cylinders Measuring Cylinders
These large capacity cylinders are made of polypropylene
  • Has double scale and hex base for stability
  • Steam autoclavable at 121 °C
  • Safer and more convenient
The handle is generously sized for large hands but with a slender enough grip for smaller hands. Hand opening is 3 1/2” x 1 1/8
The handle on the Holdfast™ graduated cylinder makes pouring safer and more convenient.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty