Would you like to log in again?
Glas-Col® STM Series Mantles, High Temperature, Chemglass
Supplier: Chemglass
Glas-Col®
CG-12500-06 CG-12500-09 CG-12500-01 CG-12500-08 CG-12500-07 CG-12500-04
80060-732EA 691.31 USD InStock
80060-732 80060-742 80060-740 80060-736 80060-734 80060-738
Glas-Col® STM Series Mantles, High Temperature, Chemglass
Heaters Heating Mantles
The Glas-Col® Series STM mantle performs the same function as a Series TM heating mantle, plus operations requiring more heat.
  • The STM mantle has a durable aluminum exterior and a resilient and pliable interior to cradle glassware
  • Withstanding an upper internal operating temperature of 650º C, the STM mantle is very resistant to burnout
  • A splash guard and poncho safety shield can be used to prevent damage from accidental spills
The higher temperature range enables operations to be performed that require more heat, such as rapid distillation of water and liquid-phase cracking in a flask, where the products are taken out of the flask through a column. Useful in distillation and fractionation processes, especially when time is important.
Caution: The STM series mantle is recommended for use with flammable solvents only when used with a poncho safety shield.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty