Would you like to log in again?
Distilling Columns Packing, Porcelain Berl Saddles, Chemglass
Supplier: Chemglass
CG-1284-02 CG-1284-01
80060-846EA 215.53 USD InStock
80060-846 80060-844
Distilling Columns Packing, Porcelain Berl Saddles, Chemglass
Distillation Column Packing
Porcelain berl saddles are ideal for vacuum distillation, absorption and extraction.
  • Saddles have an O.D of 8 mm
  • Saddles provide a high useable surface area
  • Lower gas flow resistance and good internal distribution
Packaging: Available in package of 200 and 1000 cc size boxes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty